Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202438.1 | NP_001189367.1 | 16 | 2286 | ||
NM_015608.2 | NP_056423.2 | 15 | 2184 | |||
NR_110857.1 | 16 | 2281 | ||||
NR_110858.1 | 16 | 2286 | ||||
NR_110859.1 | 16 | 2302 | ||||
XM_011539622.1 | XP_011537924.1 | 13 | 1474 | |||
XM_017016066.1 | XP_016871555.1 | 16 | 2030 | |||
GenBank mRNA | AK025270.1 | - | 4 | 257 | ||
AK091507.1 | - | 15 | 1996 | |||
AK314958.1 | - | 1 | 20 | |||
BC016903.1 | - | 4 | 282 | |||
BC026172.1 | - | 16 | 2287 | 4838438 | ||
BC105929.1 | - | 16 | 2220 | |||
BC107479.1 | - | 16 | 2225 | 40028896 |