Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015613.2 | NP_056428.1 | 1 | 105 | ||
GenBank mRNA | AB031547.1 | - | 1 | 105 |