Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025930.3 | NP_001021100.3 | 7 | 1181 | ||
NR_037162.1 | 7 | 1803 | ||||
GenBank mRNA | AK023960.1 | - | 7 | 798 | ||
AK097236.1 | - | 1 | 1419 | |||
AK125875.1 | - | 3 | 3249 | |||
AK297289.1 | - | 4 | 525 | |||
AK315140.1 | - | 5 | 571 | |||
AL096725.1 | - | 1 | 960 | |||
AL833939.1 | - | 3 | 1608 | |||
BC098298.1 | - | 1 | 428 | 40025661 | ||
BC098361.1 | - | 1 | 428 | |||
BC099735.1 | - | 1 | 428 | |||
BC105638.1 | - | 1 | 428 | |||
BP357921.1 | - | 5 | 557 | |||
CB854808.1 | - | 485 |
Gene Symbol: ARPC4-TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198793.1 | NP_001185722.1 | 8 | 1462 | ||
GenBank mRNA | BC025289.1 | - | 7 | 896 |