Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139490.1 | NP_001132962.1 | 5 | 652 | ||
NM_015650.3 | NP_056465.2 | 5 | 652 | |||
XM_006712414.2 | XP_006712477.1 | 5 | 707 | |||
XM_011510944.2 | XP_011509246.1 | 5 | 707 | |||
XM_011510945.2 | XP_011509247.1 | 5 | 707 | |||
XM_011510946.2 | XP_011509248.1 | 5 | 707 | |||
XM_011510947.2 | XP_011509249.1 | 5 | 694 | |||
XM_011510948.2 | XP_011509250.1 | 5 | 707 | |||
XM_017003789.1 | XP_016859278.1 | 5 | 707 | |||
GenBank mRNA | AF230877.1 | - | 5 | 533 | ||
AF242456.1 | - | 5 | 751 | |||
AL080153.1 | - | 4 | 390 | |||
BC059174.1 | - | 5 | 652 | 6191726 |