Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015662.2 | NP_056477.1 | 15 | 1625 | ||
XM_005264254.1 | XP_005264311.1 | 15 | 1522 | |||
XM_006711986.3 | XP_006712049.1 | 16 | 1717 | |||
XM_006711987.1 | XP_006712050.1 | 15 | 1522 | |||
XM_011532757.2 | XP_011531059.1 | 15 | 1831 | |||
XM_011532758.1 | XP_011531060.1 | 15 | 1522 | |||
XM_017003790.1 | XP_016859279.1 | 16 | 1717 | |||
XM_017003791.1 | XP_016859280.1 | 14 | 1592 | |||
XM_017003792.1 | XP_016859281.1 | 15 | 1522 | |||
XM_017003793.1 | XP_016859282.1 | 14 | 1445 | |||
XM_017003794.1 | XP_016859283.1 | 12 | 1148 | |||
XM_017003795.1 | XP_016859284.1 | 15 | 1829 | |||
GenBank mRNA | AK055826.1 | - | 15 | 1440 | ||
AK299576.1 | - | 16 | 1687 | |||
AL110218.1 | - | 12 | 1181 | |||
BC137126.1 | - | 15 | 1515 | 9021123 | ||
BC142675.1 | - | 15 | 1587 | |||
BC142729.1 | - | 15 | 1587 |