Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015666.3 | NP_056481.1 | 7 | 996 | ||
XM_005260393.1 | XP_005260450.1 | 7 | 986 | |||
XM_006723774.3 | XP_006723837.1 | 8 | 1080 | |||
XM_011528776.1 | XP_011527078.1 | 7 | 3000 | |||
XM_017027805.1 | XP_016883294.1 | 6 | 3150 | |||
XM_017027806.1 | XP_016883295.1 | 7 | 985 | |||
XM_017027807.1 | XP_016883296.1 | 6 | 3176 | |||
XM_017027808.1 | XP_016883297.1 | 7 | 1012 | |||
GenBank mRNA | AK001603.1 | - | 6 | 976 | ||
AK124572.1 | - | 7 | 3287 | |||
AK299147.1 | - | 5 | 654 | |||
AK299726.1 | - | 4 | 505 | |||
AK316027.1 | - | 6 | 776 | |||
AK316325.1 | - | 7 | 848 | |||
AL117498.1 | - | 1 | 25 | |||
BC012073.1 | - | 5 | 861 | |||
BC036716.2 | - | 6 | 970 | 4900170 |