Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006623.3 | NP_006614.2 | 11 | 1659 | ||
XM_011541226.1 | XP_011539528.1 | 13 | 1940 | |||
XM_011541227.2 | XP_011539529.1 | 13 | 1645 | |||
XM_011541228.1 | XP_011539530.1 | 13 | 1558 | |||
XM_011541231.2 | XP_011539533.1 | 13 | 1762 | |||
GenBank mRNA | AF171235.1 | - | 4 | 523 | ||
AF171236.1 | - | 8 | 1049 | |||
AF171237.1 | - | 11 | 1505 | |||
AI685723.1 | - | 2 | 359 | |||
AK093306.1 | - | 10 | 2837 | |||
AK129762.1 | - | 3 | 1409 | |||
AK309662.1 | - | 13 | 1853 | |||
AK315360.1 | - | 11 | 1580 | |||
BC000303.2 | - | 11 | 1499 | 2820369 | ||
BC001349.2 | - | 11 | 1494 | 2961389 | ||
BC011262.1 | - | 11 | 1568 | 4156703 | ||
BC023235.1 | - | 11 | 1472 | |||
BC032110.2 | - | 5 | 800 | |||
CR456795.1 | - | 11 | 1432 | |||
FJ224347.1 | - | 2124 |