Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012174.1 | NP_036306.1 | 10 | 1483 | ||
NM_153348.2 | NP_699179.2 | 10 | 1681 | |||
XM_005253867.2 | XP_005253924.1 | 9 | 1539 | |||
XM_005253868.3 | XP_005253925.1 | 9 | 1372 | |||
XM_017019175.1 | XP_016874664.1 | 10 | 1458 | |||
XM_017019176.1 | XP_016874665.1 | 11 | 1690 | |||
XM_017019178.1 | XP_016874667.1 | 6 | 2432 | |||
GenBank mRNA | AF176707.1 | - | 10 | 1458 | ||
AK289621.1 | - | 10 | 1474 | |||
BC037296.2 | - | 10 | 1681 | 4821240 |