Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318192.1 | NP_001305121.1 | 8 | 1531 | ||
NM_012450.3 | NP_036582.2 | 8 | 1528 | |||
XM_011516024.2 | XP_011514326.1 | 8 | 1311 | |||
XM_017011962.1 | XP_016867451.1 | 6 | 899 | |||
XM_017011963.1 | XP_016867452.1 | 8 | 1311 | |||
GenBank mRNA | AB208809.1 | - | 5 | 5806 | ||
AF169301.1 | - | 8 | 1501 | |||
AK313072.1 | - | 8 | 1121 | |||
BC030689.2 | - | 8 | 1510 | 4821193 |