Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012347.4 | NP_036479.1 | 9 | 1009 | ||
NM_033480.2 | NP_258441.1 | 10 | 1259 | |||
NM_033481.3 | NP_258442.2 | 10 | 1136 | |||
XM_005248995.4 | XP_005249052.1 | 10 | 943 | |||
XM_005248996.4 | XP_005249053.1 | 10 | 1094 | |||
GenBank mRNA | AB209833.1 | - | 10 | 1298 | ||
AF174597.1 | - | 6 | 598 | |||
AF176704.1 | - | 9 | 1009 | |||
AK021559.1 | - | 10 | 1295 | |||
AK095307.1 | - | 2 | 2453 | |||
AL137520.1 | - | 10 | 1259 | |||
AY858798.1 | - | 10 | 1301 | |||
BC000650.2 | - | 10 | 1136 | 3345926 | ||
BC024973.2 | - | 7 | 680 | |||
BX641082.1 | - | 2 | 1407 |