Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297437.1 | NP_001284366.1 | 2, 3 | 480 | ||
NM_012176.2 | NP_036308.1 | 2, 3 | 478 | |||
NM_033484.2 | NP_277019.1 | 2, 3 | 478 | |||
XM_011514026.2 | XP_011512328.1 | 2, 3 | 483 | |||
GenBank mRNA | AF176703.1 | - | 2, 3 | 416 | ||
AK024075.1 | - | 2, 3 | 478 | |||
AU143782.1 | - | 2, 3 | 478 | |||
BC040011.1 | - | 2, 3 | 439 | |||
BC048098.1 | - | 2, 3 | 446 | 4830192 |