Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIBCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014362.3 | NP_055177.2 | 12 | 1289 | ||
NM_198047.2 | NP_932164.1 | 12 | 1289 | |||
XM_011510953.1 | XP_011509255.1 | 12 | 1238 | |||
XM_011510954.1 | XP_011509256.1 | 7 | 689 | |||
GenBank mRNA | AI767349.1 | - | 3 | 477 | ||
AK126890.1 | - | 1 | 3137 | |||
AK222979.1 | - | 12 | 1013 | |||
AK223023.1 | - | 12 | 1013 | |||
AK308042.1 | - | 12 | 1013 | |||
AK313651.1 | - | 12 | 1019 | |||
BC005190.1 | - | 12 | 1066 | |||
BC067822.1 | - | 12 | 1018 | 5266813 | ||
CA448468.1 | - | 3 | 664 | |||
U66669.1 | - | 12 | 1042 |