Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099274.1 | NP_001092744.1 | 5 | 886 | ||
NM_012461.2 | NP_036593.2 | 5 | 886 | |||
XM_005267529.3 | XP_005267586.1 | 4 | 712 | |||
GenBank mRNA | AF195512.1 | - | 5 | 806 | ||
AK023166.1 | - | 3 | 1277 | |||
AK225379.1 | - | 5 | 830 | |||
AK294119.1 | - | 4 | 712 | |||
AK315084.1 | - | 5 | 774 | |||
BC005030.1 | - | 5 | 667 | 3690254 | ||
BC019343.1 | - | 5 | 844 | 3505009 | ||
EU851975.1 | - | 2 | 60 |