Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129981.2 | NP_001123453.1 | 3 | 615 | ||
NM_001291218.1 | NP_001278147.1 | 3 | 615 | |||
NM_001291219.1 | NP_001278148.1 | 3 | 615 | |||
NM_020349.3 | NP_065082.2 | 3 | 615 | |||
GenBank mRNA | AB058599.1 | - | 3 | 277 | ||
AJ249975.1 | - | 2 | 205 | |||
AJ304805.1 | - | 3 | 277 | |||
AJ583444.1 | - | 3 | 615 | |||
AK056990.1 | - | 3 | 557 | |||
AK314763.1 | - | 3 | 277 | |||
BC020817.1 | - | 3 | 277 | |||
BC107759.1 | - | 3 | 518 | 4096546 |