Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020184.3 | NP_064569.3 | 4 | 1902 | ||
XM_005263914.3 | XP_005263971.1 | 4 | 1819 | |||
XM_005263915.3 | XP_005263972.1 | 4 | 1819 | |||
XM_011510955.2 | XP_011509257.1 | 4 | 1819 | |||
XM_017003799.1 | XP_016859288.1 | 4 | 478 | |||
GenBank mRNA | AB046812.1 | - | 4 | 1632 | ||
AF202777.1 | - | 4 | 1663 | |||
AK022833.1 | - | 1 | 97 | |||
AK293915.1 | - | 4 | 712 | |||
BC063295.1 | - | 4 | 1902 | 30340626 |