Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013451.3 | NP_038479.1 | 41 | 4607 | ||
NM_133337.2 | NP_579899.1 | 40 | 4568 | |||
XM_005269693.4 | XP_005269750.1 | 42 | 4843 | |||
XM_005269694.4 | XP_005269751.1 | 41 | 4804 | |||
XM_011539632.2 | XP_011537934.1 | 43 | 5037 | |||
XM_017016068.1 | XP_016871557.1 | 41 | 4424 | |||
XM_017016069.1 | XP_016871558.1 | 41 | 4469 | |||
XM_017016070.1 | XP_016871559.1 | 42 | 4851 | |||
GenBank mRNA | AB033033.2 | - | 41 | 4484 | ||
AF182316.1 | - | 41 | 4580 | |||
AF207990.1 | - | 40 | 4445 | |||
AK075258.1 | - | 5 | 322 | |||
AK093890.1 | - | 1 | 588 | |||
AL096713.1 | - | 22 | 2681 |