Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018959.3 | NP_061832.2 | 5 | 585 | ||
NM_170711.2 | NP_733829.1 | 5 | 585 | |||
XM_005259531.3 | XP_005259588.1 | 5 | 535 | |||
XM_005259534.3 | XP_005259591.1 | 5 | 544 | |||
XM_005259535.3 | XP_005259592.1 | 5 | 543 | |||
XM_005259536.3 | XP_005259593.1 | 5 | 546 | |||
XM_011527904.2 | XP_011526206.1 | 5 | 1850 | |||
XM_011527905.2 | XP_011526207.1 | 5 | 1858 | |||
XM_011527906.2 | XP_011526208.1 | 5 | 1855 | |||
XM_011527907.2 | XP_011526209.1 | 5 | 1864 | |||
XM_011527908.2 | XP_011526210.1 | 5 | 1857 | |||
XM_011527909.2 | XP_011526211.1 | 5 | 1857 | |||
XM_011527910.2 | XP_011526212.1 | 5 | 937 | |||
XM_017026581.1 | XP_016882070.1 | 5 | 580 | |||
XM_017026582.1 | XP_016882071.1 | 5 | 1873 | |||
GenBank mRNA | AF181719.1 | - | 5 | 488 | ||
AK056850.1 | - | 3 | 208 | |||
AK125032.1 | - | 4 | 1881 | |||
BC012062.1 | - | 5 | 453 | 4549444 | ||
BI909277.1 | - | 5 | 569 | |||
BU557183.1 | - | 582 | ||||
KU178593.1 | - | 5 | 380 |