Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098525.2 | NP_001091995.1 | 7 | 1701 | ||
NM_001286686.1 | NP_001273615.1 | 7 | 1639 | |||
NM_018204.4 | NP_060674.3 | 7 | 1698 | |||
XM_005266344.4 | XP_005266401.1 | 7 | 1609 | |||
XM_011535043.2 | XP_011533345.1 | 6 | 1553 | |||
GenBank mRNA | AF177227.1 | - | 6 | 1393 | ||
AK001611.1 | - | 4 | 574 | |||
AK022982.1 | - | 7 | 1597 | |||
AK299083.1 | - | 7 | 1639 | |||
AL136848.1 | - | 4 | 1103 | |||
AY062261.1 | - | 7 | 1619 | |||
BC130296.1 | - | 7 | 1546 | 40146327 | ||
BC136332.1 | - | 7 | 1546 | 9020319 | ||
EF560732.1 | - | 7 | 1582 | |||
Y15758.1 | - | 7 | 1594 |