Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR7A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017506.1 | NP_059976.1 | 1 | 236 | ||
XM_011527912.2 | XP_011526214.1 | 2 | 261 | |||
XM_011527913.2 | XP_011526215.1 | 2 | 187 | |||
XM_017026583.1 | XP_016882072.1 | 2 | 150 | |||
GenBank mRNA | AK312475.1 | - | 2 | 102 | ||
BC104809.1 | - | 1 | 71 | 8143812 |