Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318014.1 | NP_001304943.1 | 4 | 1528 | ||
NM_001318022.1 | NP_001304951.1 | 4 | 1411 | |||
NM_030883.4 | NP_112145.1 | 4 | 1415 | |||
XM_011514470.2 | XP_011512772.1 | 5 | 2812 | |||
XM_011514472.2 | XP_011512774.1 | 4 | 2169 | |||
XM_011514473.2 | XP_011512775.1 | 5 | 2264 | |||
XM_011514479.2 | XP_011512781.1 | 3 | 2005 | |||
XM_011514483.2 | XP_011512785.1 | 3 | 1386 | |||
XM_011514484.2 | XP_011512786.1 | 2 | 1384 | |||
XM_017010737.1 | XP_016866226.1 | 4 | 2915 | |||
XM_017010738.1 | XP_016866227.1 | 4 | 2296 | |||
XM_017010739.1 | XP_016866228.1 | 4 | 2915 | |||
XM_017010740.1 | XP_016866229.1 | 5 | 2662 | |||
GenBank mRNA | AJ459845.1 | - | 2 | 942 | ||
AJ459846.1 | - | 2 | 955 | |||
AJ459847.1 | - | 1 | 920 | |||
AK127984.1 | - | 2 | 1629 | |||
BC048991.2 | - | 3 | 1363 | 5271584 |