Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012449.2 | NP_036581.1 | 5 | 999 | ||
GenBank mRNA | AF186249.1 | - | 5 | 864 | ||
BC011802.2 | - | 5 | 903 | 4422866 |