Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPLAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017570.4 | NP_060040.1 | 8, 9 | 1191 | ||
XM_011516960.1 | XP_011515262.1 | 8, 9 | 1467 | |||
GenBank mRNA | AB122018.1 | - | 7, 8 | 1087 | ||
AL096750.1 | - | 3, 4 | 404 | |||
BC036944.1 | - | 3, 4 | 378 | |||
BC142672.1 | - | 8, 9 | 1191 | 40148181 | ||
BC150206.1 | - | 8, 9 | 1169 | 40148883 |