Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPLAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017570.4 | NP_060040.1 | 17 | 2402 | ||
XM_011516960.1 | XP_011515262.1 | 17 | 2678 | |||
GenBank mRNA | AB122018.1 | - | 16 | 2298 | ||
AL096750.1 | - | 9 | 1387 | |||
BC036944.1 | - | 12 | 1589 | |||
BC041576.1 | - | 2 | 213 | |||
BC142672.1 | - | 17 | 2402 | 40148181 | ||
BC150206.1 | - | 17 | 2380 | 40148883 |