Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243202.1 | NP_001230131.1 | 2 | 969 | ||
NM_001243203.1 | NP_001230132.1 | 2 | 910 | |||
NM_012416.3 | NP_036548.1 | 1 | 2025 | |||
XM_017014624.1 | XP_016870113.1 | 2 | 930 | |||
GenBank mRNA | AF039023.1 | - | 1 | 1994 | ||
AK300408.1 | - | 2 | 884 | |||
AK307840.1 | - | 1 | 2000 | |||
BC098406.1 | - | 1 | 2000 | 30347107 | ||
BX537405.1 | - | 1 | 2004 | |||
DA116213.1 | - | 1 | 565 | |||
DA155272.1 | - | 1 | 216 |