Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290033.1 | NP_001276962.1 | 9 | 719 | ||
NM_012133.5 | NP_036265.3 | 9 | 719 | |||
GenBank mRNA | AB047847.1 | - | 9 | 629 | ||
AF157833.1 | - | 9 | 689 | |||
AF207598.1 | - | 9 | 685 | |||
AK002086.1 | - | 1 | 21 | |||
AK022934.1 | - | 9 | 616 | |||
AK024414.1 | - | 2 | 97 | |||
AK291409.1 | - | 9 | 709 | |||
BC017443.1 | - | 9 | 612 | |||
BC110796.1 | - | 9 | 666 | 5197024 |