Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHORDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012124.2 | NP_036256.2 | 3, 4 | 571 | ||
XM_017017540.1 | XP_016873029.1 | 3, 4 | 284 | |||
XM_017017541.1 | XP_016873030.1 | 3, 4 | 285 | |||
GenBank mRNA | AF192466.1 | - | 3, 4 | 245 | ||
AK225365.1 | - | 3, 4 | 273 | |||
AK225565.1 | - | 3, 4 | 256 | |||
AK226099.1 | - | 3, 4 | 248 | |||
AK290231.1 | - | 3, 4 | 571 | |||
AK312663.1 | - | 3, 4 | 256 | |||
BC072461.1 | - | 3, 4 | 220 | 6471725 |
Gene Symbol: LOC727896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026659.1 | 1 | 248 |