Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHORDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144073.1 | NP_001137545.1 | 3 | 632 | ||
NM_012124.2 | NP_036256.2 | 4 | 689 | |||
XM_017017540.1 | XP_016873029.1 | 4 | 402 | |||
XM_017017541.1 | XP_016873030.1 | 4 | 403 | |||
GenBank mRNA | AF123249.1 | - | 3 | 327 | ||
AF192466.1 | - | 4 | 363 | |||
AK225365.1 | - | 4 | 391 | |||
AK225565.1 | - | 4 | 374 | |||
AK226099.1 | - | 4 | 366 | |||
AK290231.1 | - | 4 | 689 | |||
AK312663.1 | - | 4 | 374 | |||
BC072461.1 | - | 4 | 338 | 6471725 |