Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012430.4 | NP_036562.2 | 3 | 405 | ||
XM_011512673.2 | XP_011510975.1 | 3 | 1921 | |||
XM_011512675.2 | XP_011510977.1 | 4 | 572 | |||
XM_011512676.2 | XP_011510978.1 | 3 | 461 | |||
GenBank mRNA | AF100749.1 | - | 3 | 387 | ||
AK057587.1 | - | 2 | 1221 | |||
AK315771.1 | - | 3 | 374 | |||
BC007122.1 | - | 2 | 353 | 3459649 | ||
BG613364.1 | - | 3 | 405 | |||
DA308910.1 | - | 3 | 405 |