Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP8L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291478.1 | NP_001278407.1 | 6, 7 | 916 | ||
NM_014371.3 | NP_055186.3 | 6, 7 | 1099 | |||
NR_111971.1 | 7, 8 | 997 | ||||
XM_005259854.3 | XP_005259911.2 | 7, 8 | 904 | |||
XM_011527916.2 | XP_011526218.1 | 6, 7 | 1006 | |||
GenBank mRNA | AB015332.1 | - | 6, 7 | 946 | ||
AB025905.1 | - | 6, 7 | 946 | |||
AB451326.1 | - | 5, 6 | 901 | |||
AB451469.1 | - | 5, 6 | 901 | |||
AF199414.1 | - | 5, 6 | 901 | |||
AJ243467.1 | - | 6, 7 | 947 | |||
AK001594.1 | - | 6, 7 | 966 | |||
AK094736.1 | - | 7, 8 | 864 | |||
AK225046.1 | - | 6, 7 | 966 | |||
AK225291.1 | - | 6, 7 | 966 | |||
AK295956.1 | - | 6, 7 | 821 | |||
AK310990.1 | - | 3, 4 | 206 | |||
AL133576.1 | - | 2, 3 | 212 | |||
BC000713.2 | - | 6, 7 | 951 | 3504388 |