Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037333.2 | NP_001032410.1 | 2 | 185 | ||
NM_001291721.1 | NP_001278650.1 | 2 | 185 | |||
NM_001291722.1 | NP_001278651.1 | 2 | 185 | |||
NM_014376.3 | NP_055191.2 | 2 | 311 | |||
XM_011534516.2 | XP_011532818.1 | 2 | 451 | |||
XM_017009341.1 | XP_016864830.1 | 2 | 300 | |||
XM_017009342.1 | XP_016864831.1 | 2 | 174 | |||
GenBank mRNA | AB032994.2 | - | 2 | 125 | ||
AF160973.1 | - | 2 | 94 | |||
AK223604.1 | - | 1 | 63 | |||
AK294220.1 | - | 2 | 142 | |||
AK295865.1 | - | 2 | 142 | |||
AK297401.1 | - | 2 | 142 | |||
AK303702.1 | - | 2 | 138 | |||
AL136549.1 | - | 2 | 184 | |||
AL559919.3 | - | 2 | 481 | |||
BC011762.1 | - | 2 | 311 | 3619680 | ||
BC021008.1 | - | 2 | 311 | |||
BG257432.1 | - | 445 | ||||
BG699081.1 | - | 2 | 190 |