Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014987.1 | NP_001014987.1 | 11 | 1033 | ||
NM_001014988.1 | NP_001014988.1 | 11 | 1030 | |||
NM_001014989.1 | NP_001014989.2 | 12 | 825 | |||
NM_014387.3 | NP_055202.1 | 10 | 1120 | |||
GenBank mRNA | AF036905.1 | - | 11 | 738 | ||
AF036906.1 | - | 10 | 846 | |||
AJ223280.1 | - | 11 | 1003 | |||
AK093815.1 | - | 3 | 2149 | |||
BC011563.1 | - | 12 | 825 | 4644259 | ||
FN432831.1 | - | 10 | 795 | |||
FN432832.1 | - | 10 | 765 | |||
FN432833.1 | - | 9 | 738 | |||
FN432834.1 | - | 10 | 652 |