Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIEXF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014388.6 | NP_055203.4 | 10, 11 | 1869 | ||
XM_011509418.2 | XP_011507720.1 | 10, 11 | 1893 | |||
GenBank mRNA | AK123674.1 | - | 4, 5 | 675 | ||
AK289613.1 | - | 10, 11 | 1849 | |||
AK314061.1 | - | 10, 11 | 1899 | |||
BC022964.1 | - | 10, 11 | 1830 | 5110673 | ||
BX648514.1 | - | 10, 11 | 1782 | |||
CR749825.1 | - | 10, 11 | 1806 |