Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014390.2 | NP_055205.2 | 19 | 2432 | ||
GenBank mRNA | AB209510.1 | - | 19 | 2370 | ||
AK096583.1 | - | 20 | 2357 | |||
AK304181.1 | - | 19 | 2324 | |||
AK309014.1 | - | 11 | 1325 | |||
AK312313.1 | - | 19 | 2386 | |||
BC017180.2 | - | 19 | 2388 | 3345037 | ||
BT009785.1 | - | 19 | 2206 | |||
U22055.1 | - | 19 | 2398 |