Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014399.3 | NP_055214.1 | 4 | 632 | ||
GenBank mRNA | AF100759.1 | - | 4 | 552 | ||
AF120265.1 | - | 4 | 458 | |||
AK026587.1 | - | 4 | 596 | |||
AK075475.1 | - | 4 | 576 | |||
AK093487.1 | - | 4 | 593 | |||
AK316595.1 | - | 4 | 575 | |||
AY358995.1 | - | 4 | 563 | |||
BC033863.1 | - | 4 | 484 | 5187772 | ||
CR542107.1 | - | 4 | 390 |