Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025247.1 | NP_001020418.1 | 4 | 704 | ||
NM_014409.3 | NP_055224.1 | 4 | 704 | |||
XM_005273099.4 | XP_005273156.1 | 4 | 657 | |||
XM_005273100.2 | XP_005273157.1 | 5 | 670 | |||
XM_011544162.1 | XP_011542464.1 | 3 | 441 | |||
XM_011544163.1 | XP_011542465.1 | 7 | 1286 | |||
XM_011544164.2 | XP_011542466.1 | 3 | 384 | |||
XM_017001045.1 | XP_016856534.1 | 6 | 1441 | |||
GenBank mRNA | AF069736.1 | - | 3 | 538 | ||
BC041094.1 | - | 4 | 627 | 5551246 |