Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014427.4 | NP_055242.1 | 10 | 1174 | ||
NM_153636.2 | NP_705900.1 | 8 | 949 | |||
XM_011523000.1 | XP_011521302.1 | 8 | 944 | |||
XM_011523001.2 | XP_011521303.1 | 8 | 709 | |||
XM_017023138.1 | XP_016878627.1 | 10 | 1138 | |||
XM_017023139.1 | XP_016878628.1 | 10 | 1149 | |||
XM_017023140.1 | XP_016878629.1 | 8 | 924 | |||
XM_017023141.1 | XP_016878630.1 | 7 | 2031 | |||
GenBank mRNA | AJ133798.1 | - | 10 | 1173 | ||
BC035334.1 | - | 8 | 887 | 5191900 | ||
BC064577.1 | - | 8 | 926 | 5727324 |