Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139318.4 | NP_647479.2 | 7 | 1182 | ||
NM_172375.2 | NP_758963.1 | 7 | 1182 | |||
GenBank mRNA | AF418206.1 | - | 7 | 1130 | ||
AF472412.1 | - | 7 | 1130 | |||
AF493798.1 | - | 7 | 1130 | |||
AK074484.1 | - | 7 | 1124 | |||
BC043409.2 | - | 7 | 1299 | |||
BC073979.1 | - | 7 | 1182 | 6199697 |