Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014429.3 | NP_055244.3 | 14 | 1312 | ||
XM_005247362.2 | XP_005247419.1 | 14 | 1269 | |||
XM_011512691.1 | XP_011510993.1 | 14 | 1275 | |||
XM_011512692.1 | XP_011510994.1 | 14 | 1275 | |||
XM_011512693.1 | XP_011510995.1 | 14 | 1275 | |||
XM_011512694.1 | XP_011510996.1 | 11 | 1026 | |||
XM_011512696.2 | XP_011510998.1 | 8 | 726 | |||
XM_011512697.2 | XP_011510999.1 | 7 | 894 | |||
XM_017006169.1 | XP_016861658.1 | 14 | 1275 | |||
XM_017006170.1 | XP_016861659.1 | 7 | 694 | |||
GenBank mRNA | AF084946.1 | - | 14 | 1287 | ||
AK302642.1 | - | 14 | 1269 | |||
AL353955.1 | - | 8 | 672 | |||
BC028680.1 | - | 14 | 1313 | |||
BC050307.1 | - | 14 | 1251 | 4838254 |