Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014429.3 | NP_055244.3 | 19 | 1976 | ||
XM_005247362.2 | XP_005247419.1 | 20 | 2023 | |||
XM_011512691.1 | XP_011510993.1 | 20 | 2029 | |||
XM_011512692.1 | XP_011510994.1 | 19 | 1939 | |||
XM_011512693.1 | XP_011510995.1 | 20 | 2029 | |||
XM_011512694.1 | XP_011510996.1 | 17 | 1780 | |||
XM_011512696.2 | XP_011510998.1 | 14 | 1480 | |||
XM_011512697.2 | XP_011510999.1 | 13 | 1648 | |||
XM_017006169.1 | XP_016861658.1 | 20 | 2029 | |||
XM_017006170.1 | XP_016861659.1 | 12 | 1358 | |||
GenBank mRNA | AF084946.1 | - | 19 | 1951 | ||
AK302642.1 | - | 18 | 1870 | |||
AL353955.1 | - | 13 | 1336 | |||
BC028680.1 | - | 19 | 1977 | |||
BC050307.1 | - | 19 | 1915 | 4838254 |