Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FILIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289987.1 | NP_001276916.1 | 6 | 1351 | ||
NM_001300866.1 | NP_001287795.1 | 5 | 1185 | |||
NM_015687.3 | NP_056502.1 | 5 | 1185 | |||
XM_005248713.3 | XP_005248770.1 | 5 | 1156 | |||
XM_005248715.4 | XP_005248772.1 | 5 | 1156 | |||
XM_011535756.2 | XP_011534058.1 | 2 | 286 | |||
GenBank mRNA | AB033101.1 | - | 3 | 573 | ||
AB086011.1 | - | 4 | 769 | |||
AK027705.1 | - | 1 | 82 | |||
AK097021.1 | - | 5 | 1025 | |||
AK290232.1 | - | 5 | 897 | |||
AL832009.2 | - | 5 | 985 | |||
BC029425.1 | - | 2 | 282 | |||
BC136443.1 | - | 5 | 940 | 9020432 | ||
BC136444.1 | - | 6 | 1106 | 9020433 | ||
BX647178.1 | - | 5 | 1136 |