Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FILIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289987.1 | NP_001276916.1 | 6 | 1697 | ||
NM_001300866.1 | NP_001287795.1 | 5 | 1531 | |||
NM_015687.3 | NP_056502.1 | 5 | 1531 | |||
XM_005248713.3 | XP_005248770.1 | 5 | 1502 | |||
XM_005248715.4 | XP_005248772.1 | 5 | 1502 | |||
XM_011535756.2 | XP_011534058.1 | 2 | 632 | |||
GenBank mRNA | AB033101.1 | - | 3 | 919 | ||
AB086011.1 | - | 4 | 1115 | |||
AK027705.1 | - | 1 | 428 | |||
AK097021.1 | - | 5 | 1371 | |||
AK290232.1 | - | 5 | 1243 | |||
AL832009.2 | - | 5 | 1331 | |||
BC029425.1 | - | 2 | 628 | |||
BC136443.1 | - | 5 | 1286 | 9020432 | ||
BC136444.1 | - | 6 | 1452 | 9020433 |