Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPAMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015692.2 | NP_056507.2 | 21 | 2724 | ||
XM_011527917.1 | XP_011526219.1 | 21 | 2691 | |||
XM_011527918.1 | XP_011526220.1 | 21 | 2727 | |||
XM_011527919.1 | XP_011526221.1 | 21 | 2727 | |||
XM_011527920.2 | XP_011526222.1 | 21 | 2727 | |||
XM_011527921.2 | XP_011526223.1 | 21 | 2727 | |||
XM_011527922.1 | XP_011526224.1 | 21 | 2727 | |||
XM_011527923.1 | XP_011526225.1 | 21 | 2727 | |||
XM_011527924.2 | XP_011526226.1 | 21 | 2727 | |||
XM_011527925.1 | XP_011526227.1 | 5 | 569 | |||
XM_017026594.1 | XP_016882083.1 | 21 | 2727 | |||
GenBank mRNA | AB033109.1 | - | 21 | 2549 | ||
AY101765.1 | - | 21 | 2568 |