Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015695.2 | NP_056510.2 | 2 | 1232 | ||
XM_005249010.2 | XP_005249067.1 | 2 | 1114 | |||
XM_005249011.3 | XP_005249068.1 | 2 | 1238 | |||
XM_011514489.1 | XP_011512791.1 | 2 | 1136 | |||
XM_011514490.1 | XP_011512792.1 | 1 | 1034 | |||
XM_011514491.1 | XP_011512793.1 | 1 | 1034 | |||
XM_011514492.1 | XP_011512794.1 | 1 | 1034 | |||
XM_017010742.1 | XP_016866231.1 | 1 | 1034 | |||
GenBank mRNA | AB033112.1 | - | 2 | 1232 | ||
BC033652.2 | - | 1 | 936 | |||
BC117387.1 | - | 1 | 1039 | 40125938 | ||
BC143917.1 | - | 1 | 1039 | 9052435 | ||
BC143918.1 | - | 1 | 1039 | 9052436 | ||
BC143919.1 | - | 1 | 1039 |