Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318865.1 | NP_001305794.1 | 3 | 437 | ||
NM_018960.5 | NP_061833.1 | 3 | 437 | |||
NR_134899.1 | 3 | 437 | ||||
GenBank mRNA | AF101477.1 | - | 3 | 433 | ||
BC032627.1 | - | 3 | 429 | 5229272 | ||
FJ224320.1 | - | 3 | 433 | |||
KU178603.1 | - | 3 | 423 | |||
KU178604.1 | - | 3 | 423 | |||
KU178605.1 | - | 3 | 423 | |||
X62250.1 | - | 3 | 571 |
Gene Symbol: CNPY3-GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318856.1 | NP_001305785.1 | 3 | 809 | ||
NM_001318857.1 | NP_001305786.1 | 2 | 681 | |||
NM_001318858.1 | NP_001305787.1 | 2 | 681 | |||
NR_134890.1 | 3 | 778 | ||||
NR_134891.1 | 2 | 681 | ||||
NR_134892.1 | 3 | 809 | ||||
GenBank mRNA | BQ947041.1 | - | 3 | 436 | ||
FY210916.1 | - | 2 | 214 | |||
FY213370.1 | - | 2 | 213 | |||
FY213777.1 | - | 2 | 214 | |||
FY213816.1 | - | 3 | 310 |