Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015697.7 | NP_056512.5 | 1, 2 | 408 | ||
XM_011531855.2 | XP_011530157.1 | 1, 2 | 619 | |||
XM_011531857.2 | XP_011530159.1 | 1, 2 | 617 | |||
XM_011531859.2 | XP_011530161.1 | 1, 2 | 616 | |||
XM_011531860.2 | XP_011530162.1 | 1, 2 | 616 | |||
XM_011531862.2 | XP_011530164.1 | 1, 2 | 615 | |||
XM_011531863.2 | XP_011530165.1 | 1, 2 | 612 | |||
XM_011531866.2 | XP_011530168.1 | 1, 2 | 608 | |||
GenBank mRNA | AJ621061.3 | - | 1, 2 | 408 | ||
AK129583.1 | - | 1, 2 | 164 | |||
AK300972.1 | - | 1, 2 | 289 | |||
BC008804.2 | - | 1, 2 | 265 | 3954787 | ||
BC020728.1 | - | 1, 2 | 305 | 4696019 | ||
BG699931.1 | - | 1, 2 | 294 |