Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BBS9 SIRNA Assays ›
Gene Symbol:
BBS9
Gene Name:
Bardet-Biedl syndrome 9
Gene Aliases:
B1, C18, D1, PTHB1
Chromosome Location:
Chr.7: 33129244 - 33637238 on Build GRCh38
UniGene ID:
Hs.372360
Species:
Human
siRNA ID
HSS120523
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BBS9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033604.1
NP_001028776.1
14, 15
1971
NM_001033605.1
NP_001028777.1
14, 15
2061
NM_014451.3
NP_055266.2
13, 14
1956
NM_198428.2
NP_940820.1
15, 16
2076
XM_005249700.3
XP_005249757.1
15, 16
2084
XM_005249701.2
XP_005249758.1
15, 16
2084
XM_011515264.2
XP_011513566.1
15, 16
2084
XM_011515265.2
XP_011513567.1
15, 16
1883
XM_011515266.2
XP_011513568.1
14, 15
2069
XM_011515267.2
XP_011513569.1
14, 15
1979
XM_011515268.1
XP_011513570.1
15, 16
2084
XM_011515269.2
XP_011513571.1
15, 16
2067
XM_011515270.2
XP_011513572.1
15, 16
2084
XM_017011990.1
XP_016867479.1
14, 15
1775
XM_017011991.1
XP_016867480.1
15, 16
1792
XM_017011992.1
XP_016867481.1
15, 16
2067
XM_017011993.1
XP_016867482.1
14, 15
1962
XM_017011994.1
XP_016867483.1
15, 16
2084
GenBank mRNA
AF095771.1
-
12, 13
1437
AK091361.1
-
2, 3
947
AK297436.1
-
12, 13
1353
AK314627.1
-
15, 16
1917
BC103831.2
-
15, 16
1696
40001794
BI254810.1
-
5, 6
560
BX647385.1
-
16, 17
2198
U85995.1
-
3, 4
191
U87408.1
-
4, 5
262
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
visual perception
protein transport
cilium assembly
fat cell differentiation
response to stimulus
protein localization to cilium
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-lg8t5:80/100.66.77.74:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline