Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033604.1 | NP_001028776.1 | 18 | 2454 | ||
NM_001033605.1 | NP_001028777.1 | 18 | 2544 | |||
NM_014451.3 | NP_055266.2 | 17 | 2439 | |||
NM_198428.2 | NP_940820.1 | 19 | 2559 | |||
XM_005249700.3 | XP_005249757.1 | 19 | 2567 | |||
XM_005249701.2 | XP_005249758.1 | 19 | 2567 | |||
XM_011515264.2 | XP_011513566.1 | 19 | 2567 | |||
XM_011515265.2 | XP_011513567.1 | 19 | 2366 | |||
XM_011515266.2 | XP_011513568.1 | 18 | 2552 | |||
XM_011515267.2 | XP_011513569.1 | 18 | 2462 | |||
XM_011515268.1 | XP_011513570.1 | 19 | 2567 | |||
XM_011515269.2 | XP_011513571.1 | 19 | 2550 | |||
XM_011515270.2 | XP_011513572.1 | 19 | 2567 | |||
XM_017011990.1 | XP_016867479.1 | 18 | 2258 | |||
XM_017011991.1 | XP_016867480.1 | 19 | 2275 | |||
XM_017011992.1 | XP_016867481.1 | 19 | 2550 | |||
XM_017011993.1 | XP_016867482.1 | 18 | 2445 | |||
GenBank mRNA | AF095770.1 | - | 2 | 286 | ||
AF095771.1 | - | 16 | 1920 | |||
AK314627.1 | - | 19 | 2400 | |||
BC103831.2 | - | 19 | 2179 | 40001794 | ||
BX647385.1 | - | 20 | 2681 | |||
U85994.1 | - | 2 | 218 | |||
U85995.1 | - | 7 | 674 | |||
U87408.1 | - | 8 | 745 |