Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127397.2 | NP_001120869.1 | 10, 11 | 1385 | ||
NM_001127398.2 | NP_001120870.1 | 9, 10 | 1223 | |||
NM_015701.4 | NP_056516.2 | 10, 11 | 1385 | |||
XM_011532766.1 | XP_011531068.1 | 10, 11 | 1299 | |||
GenBank mRNA | AF131743.1 | - | 2, 3 | 204 | ||
AF131849.1 | - | 4, 5 | 574 | |||
AK001913.1 | - | 10, 11 | 1266 | |||
AK315477.1 | - | 10, 11 | 1174 | |||
AY358717.1 | - | 9, 10 | 1107 | |||
AY453410.1 | - | 10, 11 | 1236 | |||
BC013129.1 | - | 10, 11 | 1236 | 3447840 | ||
BC022228.1 | - | 9, 10 | 1071 | 4447432 | ||
BQ017438.1 | - | 3, 4 | 308 | |||
EU794676.1 | - | 10, 11 | 1385 |