Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014470.3 | NP_055285.1 | 1 | 187 | ||
GenBank mRNA | AB040147.1 | - | 1 | 165 | ||
AF498967.1 | - | 1 | 84 | |||
AK292762.1 | - | 1 | 187 | |||
BC026356.1 | - | 1 | 187 | 4793442 | ||
Y07923.1 | - | 1 | 84 |