Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014473.2 | NP_055288.1 | 2 | 263 | ||
XM_011543323.2 | XP_011541625.1 | 2 | 301 | |||
GenBank mRNA | AF102147.1 | - | 2 | 212 | ||
AK292723.1 | - | 2 | 291 | |||
AK299480.1 | - | 2 | 253 | |||
BC002841.2 | - | 2 | 260 | 3638494 | ||
BC010874.2 | - | 2 | 224 | 3878818 | ||
BM749030.1 | - | 2 | 273 |